LaMotte Liquid Sanitizer Strength Verification Kits
Titrimetric kits verify the concentration of your sanitizer quickly and easily
  • Save money by avoiding overuse or underuse of sanitizers
Verify the strength and effectiveness of your liquid sanitizer with these colorimetric test kits. All kits contain reagents for 50 tests.
ITEM(S) ON SALE
$66.06 - $114.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test
Availability Pricing
LaMotte 4497 Liquid Sanitizer Strength Verification Kits, Chlorine Test  EW-99532-04 Mfr # 4497-01
COMPARE
10 ppm/drop Chlorine
$66.06USD / Each
$73.40USD / Each
LaMotte 7191-02 Liquid Sanitizer Strength Verification Kits, Peracetic Acid Test EW-99532-06 Mfr # 7191-02
COMPARE
50 ppm/drop peroxide; 15 ppm/drop peracetic acid Peracetic acid
$114.00USD / Each