Labconco Telescoping Base Stands
Support Labconco workstations by using one or more base stands. Total width of stands must equal width of workstation. Adjust to seven height positions to accommodate sitting or standing.
Telescoping base stands feature -3/4" tubular steel frame with glacier white powder coating; include leveling feet. They are adjustable during installation in 1" increments from 27-1/2" to 34-1/2".
$1,150.00 - $1,595.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Width (in) Hood Width (in) Description
Availability Pricing
Labconco 3746700 Telescoping Base Stand w/ Leveling Feet, 2 ft wide EW-09109-25 Mfr # 3746700
COMPARE
24 24 Telescoping Base Stand w/ Leveling Feet, 2 ft wide
$1,150.00USD / Each
Labconco 3746712 Telescoping Base Stand with Casters, 48"W X 29"D EW-09109-40 Mfr # 3746712
COMPARE
48 48 Telescoping Base Stand with Casters, 48\"W X 29\"D
$1,595.00USD / Each
Labconco 3746701 Telescoping Base Stand, 3 ft wide EW-09114-71 Mfr # 3746701
COMPARE
36 36 Telescoping Base Stand, 3 ft wide
$1,215.00USD / Each
Labconco 3746702 Telescoping Base Stand, 4 ft wide EW-09114-72 Mfr # 3746702
COMPARE
48 48 Telescoping Base Stand, 4 ft wide
$1,280.00USD / Each
Labconco 3746704 Telescoping Base Stand, 6 ft wide EW-09114-73 Mfr # 3746704
COMPARE
72 72 Telescoping Base Stand, 6 ft wide
$1,420.00USD / Each
Labconco 3746703 Telescoping Base Stand, 5 ft wide EW-09114-75 Mfr # 3746703
COMPARE
60 60 Telescoping Base Stand, 5 ft wide
$1,355.00USD / Each