Labconco® Refrigerated CentriVap® Benchtop Vacuum Concentrator
Protect your heat-sensitive samples
  • The only concentrator to combine refrigeration, -4°C, and heating, 100°C
  • Create up to nine stored programs that can be altered while in progress
  • Accommodates a variety of rotors to process up to 148 samples
  • Automatic vacuum bleed valve releases vacuum when rotor stops to prevent loss of sample
  • Models with glass lid offer protection against harsh applications
  • Choose an acid-resistant model when working with corrosive chemicals
The Labconco® refrigerated CentriVap® benchtop vacuum concentrator is the perfect unit for processing your heat-sensitive samples, such as RNA and proteins. The combination of refrigeration and heating lets you set temperatures from -4 to 100°C.
The microprocessor controls a speed of 1725 rpm, as well as the heater to provide reliable results. Recover as small as a few microliter solutes from up to 148 samples.
User-friendly keyboard allows you to store up to 9 programs to quickly run different types of samples. Convenient quick-start buttons let you initiate programs with a single push of a button. In addition, you can alter the temperature and the time parameters even while a program is in process.
Acid-resistant models feature PTFE-coated wetted materials to offer protection against corrosive chemicals and harsh applications. Optional glass lid prevents etching caused by solvents.
Order vacuum pump, CentriVap® cold trap, rotor, and sample tubes separately to meet your application needs.
$11,915.00 - $16,605.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Speed Control
Availability Pricing
Labconco 7310022 Refrigerated CentriVap® Centrifugal Vacuum Concentrator, Acid-Resistant; 115 VAC, 50/60 Hz EW-13040-63 Mfr # 7310022
COMPARE
1725 115 Fixed speed
$16,605.00USD / Each
Labconco 7310020 Refrigerated CentriVap® Centrifugal Vacuum Concentrator; 115 VAC, 50/60 Hz EW-13040-64 Mfr # 7310020
COMPARE
1725 115 Fixed speed
$11,915.00USD / Each
Labconco 7310021 Refrigerated CentriVap® Centrifugal Vacuum Concentrator, Glass Lid; 115 VAC, 50/60 Hz EW-13040-65 Mfr # 7310021
COMPARE
1725 115 Fixed speed
$13,305.00USD / Each
Labconco 7310040 Refrigerated CentriVap® Centrifugal Vacuum Concentrator; 230 VAC, 50/60 Hz EW-13040-66 Mfr # 7310040
COMPARE
1725 230 Fixed speed
$11,915.00USD / Each
Labconco 7310041 Refrigerated CentriVap® Centrifugal Vacuum Concentrator, Glass Lid; 230 VAC, 50/60 Hz EW-13040-67 Mfr # 7310041
COMPARE
1725 230 Fixed speed
$13,305.00USD / Each
Labconco 7310042 Refrigerated CentriVap® Centrifugal Vacuum Concentrator, Acid-Resistant; 230 VAC, 50/60 Hz EW-13040-68 Mfr # 7310042
COMPARE
1725 230 Fixed speed
$16,605.00USD / Each