Labconco® RapidVap® Vacuum Dry Evaporation System with RS-232
An alternative way to traditional centrifugal evaporation methods!
  • RS-232 allows convenient and easy programming of up to 9 programs
  • Durable construction prevents contamination and requires no maintenance
  • Use a combination of gentle forces – vortex motion (up to 1000 rpm), heat (ambient to 100°C) and vacuum – to process up to 110 samples
  • Easily change out sample block to accommodate a variety of tube sizes
The Labconco® RapidVap® vacuum dry evaporation system is the alternative way to traditional centrifugal evaporation methods. Faster evaporation rates are achieved through vortex action, heat, and vacuum. Heat comes directly from a 1000-watt dry block, which adds no potential source of contamination and requires no maintenance.
Microprocessor-controlled motor provides smooth variable speed of up to 1000 rpm. PTFE-coated chamber ensures that your system is protected against aggressive chemicals and harsh applications. All mechanical components are isolated from inert fumes to prolong the life of your unit.
Control the time, speed, and temperature via the easy-to-use keyboard or RS-232 communication. Up to nine programs can be set and altered while in progress. The easy-to-read LCD allows you to monitor conditions while a program is in progress. The RS-232 port is at the back of the unit for 2-way communication.
Optional lid heater provides an additional 40-watt heater to improve visibility. Heating the lid eliminates condensation caused by high boiling point solvents.
Order vacuum pump, sample block, and glassware separately to meet your application needs.
$18,005.00 - $19,815.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
Labconco 7900010 Labconco® RapidVap® Vacuum Dry Evaporation System with RS-232; 115 VAC, 50/60 Hz EW-13041-10 Mfr # 7900010
COMPARE
1000 115 Labconco® RapidVap® Vacuum Dry Evaporation System with RS-232; 115 VAC, 50/60 Hz
$18,005.00USD / Each
Labconco 7900012 Labconco® RapidVap® Vacuum Dry Evaporation System with RS-232, Lid Heater; 115 VAC, 50/60 Hz EW-13041-11 Mfr # 7900012
COMPARE
1000 115 Labconco® RapidVap® Vacuum Dry Evaporation System with RS-232, Lid Heater; 115 VAC, 50/60 Hz
$19,815.00USD / Each