Labconco® RapidVap® N2 & N2/48 Dry Evaporation System
More efficient alternative to Kuderna-Danish and rotary evaporators by offering superior evaporation rates using unique blending of gentle forces
  • Use nitrogen blow down, vortex motion, and heat to increase evaporation rate of up to 48 small samples
  • Unique design significantly reduces evaporation rate after end point is achieved
  • PTFE-coated chamber and glass lid offer protection against inert chemicals and harsh applications
  • Set up to 9 programs using user-friendly keyboard and LCD screen
The Labconco® RapidVap® N2 and N2/48 dry evaporation system is an alternative to Kuderna-Danish and rotary evaporators by combining nitrogen blow down, vortex motion, and heat to reduce the evaporation time of your sample. Nitrogen blow down decreases the partial pressure of your sample to remove solvent.
The Cool-Zone block is a unique design that significantly reduces the evaporation rate after the end point is achieved. The Cool-Zone insulates your sample and allows you to plenty of time before the samples reach dryness.
The maintenance-free microprocessor-controlled motor provides smooth variable speed of up to 500 rpm. A PTFE-coated chamber and glass lid ensure that your system is protected against aggressive chemicals and harsh applications. All mechanical components are isolated from any inert fumes and vapors that might be produced to provide optimal performance.
Control the speed, time, and temperature using the easy-to-use keyboard, while setting up to 9 programs. The LCD allows you to monitor conditions while a program is in progress as well as make any changes to the run. Optional RS-232 link features 2-way communication to set functions using the RS-232 port located at the back of the unit.
$21,865.00 - $21,965.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Power (VAC) Tubes Held
Availability Pricing
Labconco 7910010 Labconco® RapidVap® N2 Dry Evaporation System, RS-232 Link; 115 VAC, 50/60 Hz EW-13041-16 Mfr # 7910010
COMPARE
500 115 8
$21,965.00USD / Each
Labconco 7910000 Labconco® RapidVap® N2 Dry Evaporation System; 115 VAC, 50/60 Hz EW-26854-20 Mfr # 7910000
COMPARE
500 115 8
$21,865.00USD / Each