Labconco Glassware Washer Accessories, LabSolutions Powder Detergent
Removes lab contaminants such as grease, agar, blood and protein digestates
  • Rinses completely
  • Non-foaming formula does not promote leaks
LabSolutions Non-Foaming Powder Detergent is specially formulated for Labconco glassware washers. A 1% solution has a pH of approximately 12.5. Contains phosphates.
$70.50 - $2,665.00USD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Labconco FlaskScrubber 4595400 Base stand for Freestanding FlaskScrubber or SteamScrubber EW-98963-50 Mfr # 4595400
COMPARE
Base stand for Freestanding FlaskScrubber or SteamScrubber
$1,085.00USD / Each
Labconco 4591900 Drain water cooling kit, 115VAC 60Hz EW-98963-55 Mfr # 4591900
COMPARE
Drain water cooling kit, 115VAC 60Hz
$735.00USD / Each
Labconco 4588500 Lower standard rack for Glassware Washers EW-98963-61 Mfr # 4588500
COMPARE
Lower standard rack for Glassware Washers
$440.00USD / Each
Labconco FlaskScrubber 4595600 Upper Spindle Rack for FlaskScrubber and FlaskScrubber Vantage Washers EW-98963-62 Mfr # 4595600
COMPARE
Upper Spindle Rack for FlaskScrubber and FlaskScrubber Vantage Washers
$2,595.00USD / Each
Labconco 4595900 Upper Rack For Washers With Slides EW-98963-64 Mfr # 4595900
COMPARE
Upper Rack For Washers With Slides
$860.00USD / Each
Labconco 4595800 Upper Spindle Rack For Washers Without Slides EW-98963-65 Mfr # 4595800
COMPARE
Upper Spindle Rack For Washers Without Slides
$2,380.00USD / Each
Labconco 4595700 Lower Spindle Rack For Washers EW-98963-66 Mfr # 4595700
COMPARE
Lower Spindle Rack For Washers
$2,665.00USD / Each
Labconco 4596100 Mobile Conversion Kit For Washers, 115V 60hz EW-98963-80 Mfr # 4596100
COMPARE
Mobile Conversion Kit For Washers, 115V 60hz
$440.00USD / Each
Labconco 4588600 Culture Tube Insert With Lid For Washers EW-98963-85 Mfr # 4588600
COMPARE
Culture Tube Insert With Lid For Washers
$310.00USD / Each
Labconco 4589701 Petri Dish Insert For Washers EW-98963-86 Mfr # 4589701
COMPARE
Petri Dish Insert For Washers
$310.00USD / Each
Labconco 4589201 Bod Bottle Insert For Washers EW-98963-87 Mfr # 4589201
COMPARE
Bod Bottle Insert For Washers
$221.00USD / Each
Labconco 4595200 Pipet Insert For Washers, Holds 16 Pipets EW-98963-89 Mfr # 4595200
COMPARE
Pipet Insert For Washers, Holds 16 Pipets
$527.00USD / Each
Labconco 4598401 Tray Holder Insert For Washers EW-98963-90 Mfr # 4598401
COMPARE
Tray Holder Insert For Washers
$148.00USD / Each
Labconco 4424600 4" Glassware Holder For Use With Washers EW-98963-91 Mfr # 4424600
COMPARE
4" Glassware Holder For Use With Washers
$70.50USD / Each
Labconco 4424800 6" Glassware Holder For Use With Washers EW-98963-92 Mfr # 4424800
COMPARE
6" Glassware Holder For Use With Washers
$86.50USD / Each
Labconco 4542100 Bulk Tube Insert Tray For Glassware Washers EW-98963-93 Mfr # 4542100
COMPARE
Bulk Tube Insert Tray For Glassware Washers
$605.00USD / Each
Labconco 4542500 Utensil Holder For Glassware Washers EW-98963-94 Mfr # 4542500
COMPARE
Utensil Holder For Glassware Washers
$335.00USD / Each
Labconco 4422000 Labsolutions Powder Detergent, Small Pail, 140 Loads EW-98963-95 Mfr # 4422000
COMPARE
Labsolutions Powder Detergent, Small Pail, 140 Loads
$137.00USD / Each
Labconco 4422100 Labsolutions Powder Detergent, Large Pail, 400 Loads EW-98963-96 Mfr # 4422100
COMPARE
Labsolutions Powder Detergent, Large Pail, 400 Loads
$279.00USD / Each
Showing 1- 19 of 19