Labconco Fume Hood Blowers
Blowers have an epoxy-coated metal frame, fiberglass-reinforced polyester housing, and a polypropylene impeller to resist corrosion and ensure a long service life.
$4,740.00 - $5,620.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Blower Type Exhaust Volume (CFM) Power (VAC)
Availability Pricing
Labconco 7181000 Standard air blower, 1/4 hp, 115 VAC/60 Hz EW-09078-10 Mfr # 7181000
COMPARE
Standard 720 115
$4,740.00USD / Each
Labconco 7181100 Explosion-proof air blower, 1/4 hp, 960 cfm, 115 VAC/60 Hz EW-09078-60 Mfr # 7181100
COMPARE
Explosion-proof 960 115
$5,620.00USD / Each
Labconco 7180300 Explosion-proof air blower, 1/4 hp, 700 cfm, 115 VAC/60 Hz EW-09078-90 Mfr # 7180300
COMPARE
Explosion-proof 700 115
$5,040.00USD / Each