Labconco CentriVap Micro IR Vacuum Centrifugal Concentrators
Easily transport this compact comprehensive vacuum concentrator from lab to lab!
  • Heated, transparent glass lid allows you to view your samples while they are spinning
  • Tiny footprint under 1 cubic foot saves valuable benchspace
  • Magnetic induction pump runs quietly with minimal vibration
More about the CentriVap Micro IR Vacuum ConcentratorThe CentriVap micro IR vacuum concentrator was built to suit your sample preparation needs with the smallest footprint available on the market. Remove solvent from your samples using vacuum, centrifugal force, and gentle heat transfer. The centrifugation eliminates bumping and foaming as vacuum is applied, and also concentrates the sample.
The PTFE-coated aluminum chamber ensures excellent chemical resistance and longevity of your unit. Monitor your samples with the micro IR strobe light while the rotor is still turning - no need to vent your chamber! The PTFE-coated diaphragm pump offers robust chemical resistance and is oil-free for low maintenance.
The infrared (IR) -emitting lid serves two primary functions: it prevents condensation and preserves your samples by eliminating cross-contamination, in addition to allowing direct heat to samples, speeding up evaporation time. An additional microprocessor-controlled, 300-watt heater is provided for samples that may be sensitive to radiant heat. Use to preheat your chamber or deliver controlled heat settings. Easy-to-read LCD displays actual temperature during "standby" and "run" mode. Standby light indicates when the selected heater is on and running.
$10,250.00 - $10,255.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrical Plug Type Power (VAC)
Availability Pricing
Labconco 7701020 CentriVap Micro IR Vacuum Centrifugal Concentrator; 115V US EW-13040-11 Mfr # 7701020
COMPARE
US 115
$10,255.00USD / Each
Labconco 7701037 CentriVap Micro IR Vacuum Centrifugal Concentrator; 230V China/Australia EW-13040-17 Mfr # 7701037
COMPARE
China/Australia 230
$10,250.00USD / Each