Labconco 47- and 70-Inch Basic Fume Hoods
Perfect when you need a full-sized ventilated hood to handle small quantities of chemicals
Benefit from renowned Labconco design and quality—while staying within a limited budget. Basic™ hoods enable safe light-duty chemical handling for a broad range of applications. These hoods utilize by-pass airflow design to assure safe continuous ventilation regardless of sash height. All models incorporate advanced design features such as a removable curved air foil and an adjustable two-piece baffle to ensure optimal air routing. Applications should limit long-term exposure to harsh solvents and highly-corrosive vapors.
All fume hoods include a 3/16" (0.5 cm) thick tempered safety glass counterbalanced sash and a 10" (25.4 cm) diameter duct collar. Hoods are constructed of glacier white epoxy-coated 16 gauge steel and include a removable front panel and a 4" (10.2 cm) L x 41/2" (11.4 cm) W utility pass-through port with molded neoprene cover. Hoods do not include a cabinet or work surface—order in accessories.
Models are available with or without a built-in blower, and in standard or spark-proof configurations. Blowers can be mounted any of three ways—within the hood, above the hood, or on the roof of your building.
The 115 VAC models are shipped fully assembled; 220 VAC models require assembly. Cord and plug are not included.
ITEM(S) ON SALE
$5,472.00 - $9,472.50USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Power (VAC) Description
Availability Pricing
Labconco 2247400 47" Basic Fume Hood, with no blower, 115 VAC/60 Hz EW-33503-00 Mfr # 2247400
COMPARE
48 115 47\" Basic Fume Hood, with no blower, 115 VAC/60 Hz
$5,472.00USD / Each
$6,080.00USD / Each
Labconco 2247300 47" Basic Fume Hood, with standard blower; 115 VAC/60 Hz EW-33503-10 Mfr # 2247300
COMPARE
53 115 47\" Basic Fume Hood, with standard blower; 115 VAC/60 Hz
$7,060.50USD / Each
$7,845.00USD / Each
Labconco 2246604 47" Basic Fume Hood, with standard blower; 220 VAC/50 Hz EW-33503-15 Mfr # 2246604
COMPARE
53 220 47\" Basic Fume Hood, with standard blower; 220 VAC/50 Hz
$7,060.50USD / Each
$7,845.00USD / Each
Labconco 2247500 47" Basic Fume Hood, with explosion-proof blower, 115 VAC/60 Hz EW-33503-20 Mfr # 2247500
COMPARE
53 115 47\" Basic Fume Hood, with explosion-proof blower, 115 VAC/60 Hz
$8,433.00USD / Each
$9,370.00USD / Each
Labconco 2246300 70" Basic Fume Hood, with standard blower; 115 VAC/60 Hz EW-33504-10 Mfr # 2246300
COMPARE
53 115 70\" Basic Fume Hood, with standard blower; 115 VAC/60 Hz
$9,279.00USD / Each
$10,310.00USD / Each
Labconco 2246704 70" Basic Fume Hood, with standard blower; 220 VAC/50 Hz EW-33504-15 Mfr # 2246704
COMPARE
53 220 70\" Basic Fume Hood, with standard blower; 220 VAC/50 Hz
$9,472.50USD / Each
$10,525.00USD / Each