LabChem Water, Deionized, ASTM Type I

Quality and consistency your lab can count on
  • Water meets ACS Reagent grade ASTM Type I standards
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$35.90 - $530.50USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Packaging Volume (mL) Description
Availability Pricing
LabChem Water, Deionized, ACS Grade, ASTM Type I, 1 L EW-80054-96 Mfr # LC267402
COMPARE
Poly Bottle 1000 Water, Deionized, ACS Grade, ASTM Type I; 1 L
$35.90USD / Each
LabChem Water, Deionized, ACS Grade, ASTM Type I, 200 L EW-80054-98 Mfr # LC26740200
COMPARE
Poly Drum 200000 Water, Deionized, ACS Grade, ASTM Type I; 200 L
$530.50USD / Each
LabChem Water, Deionized, ACS Grade, ASTM Type I, 4 L EW-80054-99 Mfr # LC267404
COMPARE
Poly Bottle 4000 Water, Deionized, ACS Grade, ASTM Type I; 4 L
$45.90USD / Each
LabChem Water, Deionized, ACS Grade, ASTM Type I, 20 L EW-80055-01 Mfr # LC267405
COMPARE
Cubitainer 20000 Water, Deionized, ACS Grade, ASTM Type I; 20 L
$108.00USD / Each
LabChem Water, Deionized, ACS Grade, ASTM Type I, 10 L EW-80055-02 Mfr # LC267406
COMPARE
Cubitainer 10000 Water, Deionized, ACS Grade, ASTM Type I; 10 L
$77.40USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat