Lab Companion Reciprocal Shaking Water Baths
PID temperature and speed control at economical prices
  • Gentle start shaking minimzes shock to samples
  • Dual LED displays show actual or set temperature and shaking speed
  • Included gable cover minimizes fluid loss from evaporation
These shaking baths feature microprocessor temperature control with built-in calibration. Store up to three temperature settings for fast recall. Shaker mechanism has its own control key enabling you to use the unit as a constant temperature bath. Digital timer can be set from 1 to 99 hr 59 min and includes a delayed on/off capability. Shaking carriage can be removed allowing you to attach racks without contacting the fluid. Safety features include independent overtemperature and low-level alarms.
$3,700.00 - $5,040.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L)
Availability Pricing
Jeio Tech Lab Companion AAH44063U 17L Reciprocal Shaking Water Bath - 120VAC, 60Hz EW-12054-00 Mfr # AAH44063U
COMPARE
115 17
$3,700.00USD / Each
Jeio Tech Lab Companion AAH44062K 17L Reciprocal Shaking Water Bath - 230VAC, 50Hz EW-12054-05 Mfr # AAH44062K
COMPARE
230 17
$3,700.00USD / Each
Jeio Tech Lab Companion AAH44113U 25L Reciprocal Shaking Water Bath - 120VAC, 60Hz EW-12054-10 Mfr # AAH44113U
COMPARE
115 25
$4,095.00USD / Each
Jeio Tech Lab Companion AAH44311K 55L Reciprocal Shaking Water Bath - 230VAC, 60Hz EW-12054-30 Mfr # AAH44311K
COMPARE
230 55
$5,040.00USD / Each
Jeio Tech Lab Companion AAH44312K 55L Reciprocal Shaking Water Bath - 230VAC, 50Hz EW-12054-35 Mfr # AAH44312K
COMPARE
230 55
$5,040.00USD / Each