Lab Companion Multiposition Stirring Hot Plates
Simultaneous stirring at multiple positions with stable heating.
  • Up to 15 independently controlled stirring positions on one heated top plate
  • Ideal for multiple sample testing under the same condition
These multiposition stirring hot plates have a smooth-start mechanism that prevents decoupling of stir bars even with viscous media. Featuring an easy-to-read and intuitive control panel with bright LED lights indicating stirring position and a heating operation light. Select from four operating modes: all, row, column, or individual. Control each mode with the built in timer for unattended operation. Over-temperature and current limit protection provide added user safety.
Integrated heater and aluminum top plate design provide exceptional heat up and transfer to samples. Powder-coated steel body design protects internal components from splashes and spills and attached non-slip silicone top plate mat prevents unintended slips.
$4,794.00 - $4,924.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Power (VAC)
Availability Pricing
Jeio Tech Lab Companion AAHK34013U 9 Position Stirring Hot Plate, 120 V. EW-51460-06 Mfr # AAHK34013U
COMPARE
9 120
$4,794.00USD / Each
Jeio Tech Lab Companion AAHK34015K 9 Position Stirring Hot Plate, 230 V. EW-51460-07 Mfr # AAHK34015K
COMPARE
9 230
$4,924.00USD / Each