Koflo PVC Mixers Without Injection Port
Mix fluids as you pump them through a pipe line
  • Economical, long-lasting, and more convenient than most motorized mixers
  • In-line mixers prevent overmixing and undermixing, and resist fouling in your pipes or tubes
  • Virtually maintenance-free and need no spare parts
In-line mixers are more convenient than most motorized mixers. Instead of mixing fluids with a motorized mixer before pumping, you can mix fluids while you pump them through a pipe line. In-line mixers prevent overmixing and undermixing, and resist fouling in your pipes or tubes. They are virtually maintenance free and need no spare parts.
In-line mixers provide efficient mixing and can withstand large pressure changes — up to 1500 psi with smallest sizes. Mixing elements in pipe and tube mixers are securely attached to the mixer walls, so internal pressure changes cannot collapse elements and stop flow.
These mixers are designed for blending light-viscosity treatment chemicals into a water stream. Each mixer is constructed of schedule 80 PVC and has four fixed, solid PVC mixing elements. Maximum working pressure is 150 psi. Connections are 150-lb raised face flanged ends.
Note: these mixers are designed for high flow rates. They use four elements instead of six to minimize pressure loss.
$353.00 - $1,660.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 1.5-80-4-4-9 PVC static mixer without injection port; 1-1/2" dia EW-04668-63 Mfr # 1.5-80-4-4-9
COMPARE
10 150 lb raised-face flange 4
$353.00USD / Each
Koflo 2-80-4-4-9 PVC static mixer without injection port; 2" dia EW-04668-65 Mfr # 2-80-4-4-9
COMPARE
12 150 lb raised-face flange 4
$437.00USD / Each
Koflo 2.5-80-4-4-9 PVC static mixer without injection port; 2-1/2" dia EW-04668-67 Mfr # 2.5-80-4-4-9
COMPARE
14 150 lb raised-face flange 4
$708.00USD / Each
Koflo 3-80-4-4-9 PVC static mixer without injection port; 3" dia EW-04668-69 Mfr # 3-80-4-4-9
COMPARE
18 150 lb raised-face flange 4
$824.00USD / Each
Koflo 4-80-4-4-9 PVC static mixer without injection port; 4" dia EW-04668-71 Mfr # 4-80-4-4-9
COMPARE
23 150 lb raised-face flange 4
$1,195.00USD / Each
Koflo 6-80-4-4-9 PVC static mixer without injection port; 6" dia EW-04668-73 Mfr # 6-80-4-4-9
COMPARE
33 150 lb raised-face flange 4
$1,660.00USD / Each