Koflo PFA Schedule 80 In-Line Pipe Mixer

Mix fluids as you pump them through a pipe line
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
$568.00 - $976.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 1/2-80-8P-6-2 Pipe Mixer, PFA schedule 80, 1/2" NPT(M), 6 elements EW-04668-20 Mfr # 1/2-80-8P-6-2
COMPARE
5 0.5\" NPT(M) 6
$568.00USD / Each
Koflo 1/2-80-8P-12-2 Pipe Mixer, PFA schedule 80, 1/2" NPT(M), 12 elements EW-04668-22 Mfr # 1/2-80-8P-12-2
COMPARE
9 0.5\" NPT(M) 12
$976.00USD / Each
Koflo 3/4-80-8P-6-2 Pipe Mixer, PFA schedule 80, 3/4" NPT(M), 6 elements EW-04668-24 Mfr # 3/4-80-8P-6-2
COMPARE
6 0.75\" NPT(M) 6
$590.00USD / Each
Koflo 1-80-8P-6-2 Pipe Mixer, PFA schedule 80, 1" NPT(M), 6 elements EW-04668-27 Mfr # 1-80-8P-6-2
COMPARE
8 1\" NPT(M) 6
$672.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat