Kobold Industrial Flow Indicators with Plastic Rotor
Integral cleaning mechanism ensures clear, unobstructed viewing
  • Transparent glass cylinder
  • Self cleaning mechanism for the glass cylinder
  • Universal mountability
With a simple twist of the sight glass, integral wipers clean the viewing area to allow unobstructed observation of th rotor. The offending contaminants are then simply washed away by the medium flow. This process may be repeated as often as desired - no fuss - no downtime.
Available with brass or stainless steel fittings with female threads 1/4" to 1-1/2" NPT. The body is made of a hard glass (Duran) and the rotor and wipers are synthetic. The device is kept tight and rotatable through the use of low friction o-rings.
$213.30 - $679.50USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Max Flow Rate (GPM) Process Connection
Availability Pricing
Kobold Industrial Flow Indicator, Brass Fittings, 6.34 GPM EW-32481-61 Mfr # DAA-5115
COMPARE
Brass 6.34 1/2" NPT
$290.00USD / Each
Kobold Industrial Flow Indicator, Brass Fittings, 2.11 GPM EW-32481-62 Mfr # DAA5108
COMPARE
Brass 2.11 1/4"NPT
$213.30USD / Each
Kobold Industrial Flow Indicator, Brass Fittings, 3.17 GPM EW-32481-63 Mfr # DAA5110
COMPARE
Brass 3.17 3/8"NPT
$247.90USD / Each
Kobold Industrial Flow Indicator, Brass Fittings, 12.15 GPM EW-32481-64 Mfr # DAA5120
COMPARE
Brass 12.15 3/4" NPT
$311.10USD / Each
Kobold Industrial Flow Indicator, Brass Fittings, 17.44 GPM EW-32481-65 Mfr # DAA5125
COMPARE
Brass 17.44 1" NPT
$330.50USD / Each
Kobold Industrial Flow Indicator, Brass Fittings, 29.06 GPM EW-32481-66 Mfr # DAA5132
COMPARE
Brass 29.06 1 1/4"NPT
$412.10USD / Each
Kobold Industrial Flow Indicator, Stainless Steel Fittings, 2.11 GPM EW-32481-68 Mfr # DAA5208
COMPARE
Stainless Steel 2.11 1/4"NPT
$324.40USD / Each
Kobold Industrial Flow Indicator, Stainless Steel Fittings, 3.17 GPM EW-32481-69 Mfr # DAA5210
COMPARE
Stainless Steel 3.17 3/8"NPT
$366.20USD / Each
Kobold Industrial Flow Indicator, Stainless Steel Fittings, 6.34 GPM EW-32481-70 Mfr # DAA5215
COMPARE
Stainless Steel 6.34 1/2" NPT
$401.60USD / Each
Kobold Industrial Flow Indicator, Stainless Steel Fittings, without Rotor, 3.17 GPM EW-32481-82 Mfr # DAA6208
COMPARE
Stainless Steel 3.17 1/4"NPT
$312.20USD / Each
Kobold Industrial Flow Indicator, Stainless Steel Fittings, without Rotor, 23.78 GPM EW-32481-86 Mfr # DAA6225
COMPARE
Stainless Steel 23.78 1" NPT
$546.50USD / Each
Kobold Industrial Flow Indicator, Stainless Steel Fittings, without Rotor, 39.63 GPM EW-32481-87 Mfr # DAA6232
COMPARE
Stainless Steel 39.63 1 1/4"NPT
$679.50USD / Each
Showing 1- 12 of 12