Cole-Parmer® TELOS® SAX Ion Exchange SPE Columns
Extract acidic analytes with a negative charge from aqueous samples
  • Features Quaternary amine sorbent
TELOS SAX (quaternary amine) columns have average pore diameter of 70 Å with 0.4 meq/g exchange capacity. All columns are made of polypropylene and come with Luer-slip outlet. Ionizable acidic analytes with pK range 2 to 6.
$141.00 - $264.00USD / Pkg of 30
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
6 1000
$229.00USD / Pkg of 30
COMPARE
1 50
$218.00USD / Pkg of 100
COMPARE
1 100
$236.00USD / Pkg of 100
COMPARE
3 100
$212.00USD / Pkg of 50
COMPARE
3 200
$141.00USD / Pkg of 50
COMPARE
3 500
$264.00USD / Pkg of 50