Cole-Parmer® TELOS® NH2 Flash Chromatography Columns with Luer-lok® inlet and outlet
Use with SP and Idolera™ systems from Biotage®
  • Optimized column geometry provides maximum recovery
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
Make connections to flash chromatography instruments quick, easy, and secure with Luer-lok connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.
$756.00USD / Pkg of 20
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g)
Availability Pricing
COMPARE
20
$756.00USD / Pkg of 20