Cole-Parmer® Screw Caps for 9 mm Short Thread Vials
No more damaged autosampler needles or sample loss
  • Ensure septa is not deformed due to overtightening—extends the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond® septum to avoid sample contamination
Short thread polypropylene caps fit short thread vials. Caps are available in different colors, septum materials, and open or closed configurations to suit your application. UltraBond cap and septum form a one-piece unit using a bonding process without adhesives for contamination-free results. Pre-slit septa prevent vacuum forming inside the vial.
$144.00 - $671.00USD / Pkg of 1000
43 variations of this product are available.
See All Product Options Below
43 PRODUCT OPTIONS
Compareitem specifications
Description Cap color Septum Type
Availability Pricing
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, UltraBond Silicone/Red PTFE Septa; 1000/pk Blue UltraBond Silicone/Red PTFE
$211.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, Red PTFE/Silicone/Red PTFE Septa; 1000/pk Blue Red PTFE/Silicone/Red PTFE
$222.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Dark Blue Polypropylene, Silicone/Red PTFE Septa; 1000/pk Dark Blue Silicone/Red PTFE
$146.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, Pre-Slit Silicone/Blue PTFE Septa; 1000/pk Blue Pre-Slit Silicone/Blue PTFE
$231.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, Red Silicone/White PTFE Septa; 1000/pk Blue Red Silicone/White PTFE
$144.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, Pre-Slit Silicone/Red PTFE Septa; 1000/pk Blue Pre-Slit Silicone/Red PTFE
$162.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Dark Blue Polypropylene, UltraBond Silicone/Red PTFE Septa; 1000/pk Dark Blue UltraBond Silicone/Red PTFE
$204.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Dark Blue Polypropylene, Pre-Slit UltraBond Silicone/Red PTFE Septa; 1000/pk Dark Blue Pre-Slit UltraBond Silicone/Red PTFE
$257.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Green Polypropylene, Silicone/Red PTFE Septa; 1000/pk Green Silicone/Red PTFE
$146.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Red Polypropylene, Silicone/Red PTFE Septa; 1000/pk Red Silicone/Red PTFE
$146.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Transparent Polypropylene, PTFE Septa; 1000/pk Transparent PTFE
$212.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, PTFE Septa; 1000/pk Blue PTFE
$212.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Closed Blue Polypropylene, PTFE Septa; 1000/pk Blue PTFE
$222.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Transparent Polypropylene, Rubber/TEF Septa; 1000/pk Transparent Rubber/TEF
$235.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Transparent Polypropylene, Red Rubber/PTFE Septa; 1000/pk Transparent Red Rubber/PTFE
$264.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, Rubber/TEF Septa; 1000/pk Blue Rubber/TEF
$250.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Blue Polypropylene, Red Rubber/PTFE Septa; 1000/pk Blue Red Rubber/PTFE
$264.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Closed Blue Polypropylene, Rubber/PTFE Septa; 1000/pk Blue Rubber/PTFE
$300.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Green Polypropylene, Rubber/TEF Septa; 1000/pk Green Rubber/TEF
$275.00USD / Pkg of 1000
COMPARE
Short Thread Cap, 9mm, Open Red Polypropylene, Rubber/TEF Septa; 1000/pk Red Rubber/TEF
$275.00USD / Pkg of 1000
Showing 1 - 20 of 43