Kinesis KX Mixed Cellulose Ester (MCE) Filter Membranes
Remove particulate without adversely affecting your final product
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results
$59.20 - $170.75USD / Pkg of 100
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
COMPARE
13 Mixed Cellulose Ester 0.22
$59.20USD / Pkg of 100
COMPARE
13 Mixed Cellulose Ester 0.45
$59.20USD / Pkg of 100
COMPARE
25 Mixed Cellulose Ester 0.22
$84.80USD / Pkg of 100
COMPARE
25 Mixed Cellulose Ester 0.45
$84.80USD / Pkg of 100
COMPARE
47 Mixed Cellulose Ester 0.22
$152.00USD / Pkg of 100
COMPARE
47 Mixed Cellulose Ester 0.45
$170.75USD / Pkg of 100