Cole-Parmer® KX+ EPA (VOA) Vials
Specifically designed for use with volatile organic compounds
  • Made of chemically inert Type I borosilicate glass
EPA (VOA) vials meet or exceed EPA standards for volatile organic analysis. Clear or amber vials are available in 20, 40, and 60 mL sizes. Order polypropylene screw caps or SURE-Clean™ caps separately.
$485.00 - $969.00USD / Pkg of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Tube Color Volume (mL)
Availability Pricing
COMPARE
57 Clear 20
$485.00USD / Pkg of 1000
COMPARE
95 Amber 40
$850.00USD / Pkg of 1000
COMPARE
140 Amber 60
$969.00USD / Pkg of 1000