Kinesis Filtration Systems
All-glass filtration system, filter holder with funnel, flask, and aluminum clamp
  • Minimize contact between sample and filtration system
  • Complete filtration assembly just add a membrane
Use our all-glass filtration systems for your work with particulate, microbiological, and HPLC to minimize contact between material and sample. 47 mm glass membrane funnel mounts on a flask reservoir with a ground glass joint. The glass membrane funnel’s drip tube is positioned below the ¼” barb to stop any sample aspiration in to the vacuum hose. Clean the all-glass system with organic and aqueous solvents.
$251.50 - $356.00USD / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Funnel Size (mL) Receiver Capacity (mL)
Availability Pricing
COMPARE
47 300 1000
$251.50USD / Pkg of 1
COMPARE
47 500 2000
$356.00USD / Pkg of 1