Justrite Drum Lock Sets
Safely secure drum contents without the risk of contamination
  • Install in just seconds
  • Fits most popular styles of steel and plastic 55-gallon drums
  • Each set contains two locks
Justrite drum locks are a cost-effective solution to controlling storage concerns of all types of liquids, semi-liquids, and powders that come in standard drums. These drum locks fit the most popular style of steel and plastic 55-gallon (200L) drums. The locks are corrosion resistant, non-sparking zinc die-cast. They install over bung flange — no potential for content contamination.
$111.75 - $121.75USD / Set of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Justrite 8508 Steel Drum Lock Set EW-81792-61 Mfr # 08508
COMPARE
Steel Drum Lock Set
$121.75USD / Set of 1
Justrite 8509 Plastic Drum Lock Set EW-81792-65 Mfr # 08509
COMPARE
Plastic Drum Lock Set
$111.75USD / Set of 1