Jun-Air® Oilless Rocking-Piston Air Compressors
Clean, quiet, and reliable
  • Tanks are internally powder-coated to eliminate corrosion from contaminated air
  • Low noise and vibration levels allow installation at point of use
  • Continuous duty cycle and a minimum of 8000 hours operation ensure clean, quiet air is always available
These oil-free air compressors provide air power suitable for laboratories and other environments with delicate machinery and critical instrumentation. Choose a model with a protective metal cabinet housing to reduce the sound level even further than the base model's already quiet level. For applications requiring dry air, such as those sensitive to microbial growth, choose from the models with the integrated adsorbent dryer.
$2,470.00 - $6,520.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Power (VAC) Tank Size (Gal)
Availability Pricing
JUN-AIR Jun-Air Oilless Piston Air Compressor, 1.4 cfm, 120 psi, 120 V EW-16202-10 Mfr # 1760428
COMPARE
1.41 120 1.1
$2,470.00USD / Each
JUN-AIR Jun-Air Oilless Piston Air Compressor, 3.2 cfm, 120 psi, 4 L tank, 120 V EW-16202-12 Mfr # 1760408
COMPARE
3.2 120 1.1
$2,845.00USD / Each
JUN-AIR Jun-Air Oilless Piston Air Compressor, 3.2 cfm, 120 psi, 25 L tank, 120 V EW-16202-14 Mfr # 1760418
COMPARE
3.2 120 6.6
$3,270.00USD / Each
JUN-AIR Jun-Air Oilless Piston Air Compressor w/ Dryer & Sound Proof Cabinet, 4 L, 120V EW-16202-24 Mfr # 1770008
COMPARE
2.75 120 1.1
$5,425.00USD / Each
JUN-AIR Jun-Air Oilless Piston Air Compressor w/ Dryer & Sound Proof Cabinet, 4 L, 230V EW-16202-25 Mfr # 1770009
COMPARE
2.75 230 1.1
$6,520.00USD / Each