Julabo Ultra-Low Refrigerated-Heating Circulators
Ultra-low temperature circulators with short cool down times
  • Low-level audible and optical alarms protect your process and the unit
  • Suitable for external temperature applications with simultaneous operation in the bath tank
  • Improved insulation prevents ice formation
  • Ideal for use with flammable bath fluids —Class III (FL) approval
  • Available with RS-232 or RS-232 and RS-485 interface for PC connection
These circulating baths are designed to achieve low temperatures and maintain excellent temperature stability for consistent results. Powerful circulating pumps along with high heating and cooling capacities allow for quick heat-up and cool-down times. The smart pump technology offers consistent pump capacity with the electronically adjustable pump stages. Ensure maximum safety with adjustable high-temperature cutoff, dry running protection, and low liquid level warnings with both optical and audible alarms. These recirculators feature a removable grid for effortless condenser cleaning to keep your chiller running at full cooling capacity. The FP models all feature energy-saving cooling control, which can save you up to 90% energy in comparison to non-FP models.
$17,669.00 - $66,489.00USD / Each
34 variations of this product are available.
See All Product Options Below
34 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Reservoir Capacity (L) Description
Availability Pricing
Julabo FP89-HL 8L Refrigerated/Heating Circulator, 100°C; 230V/50Hz EW-12151-72 Mfr # 9312689.03
COMPARE
-90 8 8L Refrigerated/Heating Circulator, 100°C; 230V/50Hz
$32,354.00USD / Each
Julabo FP51-SL 11L Refrigerated/Heating Circulator, 200°C; 208V/60Hz EW-12151-73 Mfr # 9352751.16.N
COMPARE
-51 11 11L Refrigerated/Heating Circulator, 200°C; 208V/60Hz
$17,669.00USD / Each
Julabo FP51-SL 11L Refrigerated/Heating Circulator, 200°C; 230V/50Hz EW-12151-74 Mfr # 9352751.06
COMPARE
-51 11 11L Refrigerated/Heating Circulator, 200°C; 230V/50Hz
$19,082.00USD / Each
Julabo FP52-SL 24L Refrigerated/Heating Circulator, 100°C; 230V/60Hz EW-12151-75 Mfr # 9352752.16.N
COMPARE
-60 24 24L Refrigerated/Heating Circulator, 100°C; 230V/60Hz
$35,839.00USD / Each
Julabo FP52-SL 24L Refrigerated/Heating Circulator, 100°C; 400V/50Hz EW-12151-76 Mfr # 9352752.07.N
COMPARE
-60 24 24L Refrigerated/Heating Circulator, 100°C; 400V/50Hz
$38,706.00USD / Each
Julabo FP52-SL 24L Refrigerated/Heating Circulator, 150°C; 400V/50Hz EW-12151-80 Mfr # 9352752.07.N150
COMPARE
-60 24 24L Refrigerated/Heating Circulator, 150°C; 400V/50Hz
$43,053.00USD / Each
Julabo FPW55-SL 27L Water-Cooled Refrigerated/Heating Circulator, 100°C; 230V/60Hz EW-12151-85 Mfr # 9352756.16.N
COMPARE
-60 27 27L Water-Cooled Refrigerated/Heating Circulator, 100°C; 230V/60Hz
$41,885.00USD / Each
Julabo FPW52-SL 24L Water-Cooled Refrigerated/Heating Circulator, 150°C; 230V/60Hz EW-12151-87 Mfr # 9352753.16.N150
COMPARE
-60 24 24L Water-Cooled Refrigerated/Heating Circulator, 150°C; 230V/60Hz
$40,136.00USD / Each
Julabo FPW52-SL 24L Water-Cooled Refrigerated/Heating Circulator, 150°C; 400V/50Hz EW-12151-88 Mfr # 9352753.07.N150
COMPARE
-60 24 24L Water-Cooled Refrigerated/Heating Circulator, 150°C; 400V/50Hz
$43,347.00USD / Each
Julabo FPW55-SL 27L Water-Cooled Refrigerated/Heating Circulator, 150°C; 400V/50Hz EW-12151-90 Mfr # 9352756.07.N150
COMPARE
-60 27 27L Water-Cooled Refrigerated/Heating Circulator, 150°C; 400V/50Hz
$48,376.00USD / Each
Julabo FP90-SL 22L Refrigerated/Heating Circulator, 100°C; 400V/50Hz EW-12151-92 Mfr # 9352790.07.N
COMPARE
-90 22 22L Refrigerated/Heating Circulator, 100°C; 400V/50Hz
$52,093.00USD / Each
Julabo FP90-SL 22L Refrigerated/Heating Circulator, 150°C; 230V/60Hz EW-12151-93 Mfr # 9352790.16.N150
COMPARE
-90 22 22L Refrigerated/Heating Circulator, 150°C; 230V/60Hz
$51,636.00USD / Each
Julabo FP90-SL 22L Refrigerated/Heating Circulator, 150°C; 400V/50Hz EW-12151-94 Mfr # 9352790.07.N150
COMPARE
-90 22 22L Refrigerated/Heating Circulator, 150°C; 400V/50Hz
$55,767.00USD / Each
Julabo FPW90-SL 22L Water-Cooled Refrigerated/Heating Circulator, 100°C; 230V/60Hz EW-12151-95 Mfr # 9352791.16.N
COMPARE
-90 22 22L Water-Cooled Refrigerated/Heating Circulator, 100°C; 230V/60Hz
$48,508.00USD / Each
Julabo FPW90-SL 22L Water-Cooled Refrigerated/Heating Circulator, 100°C; 400V/50Hz EW-12151-96 Mfr # 9352791.07.N
COMPARE
-90 22 22L Water-Cooled Refrigerated/Heating Circulator, 100°C; 400V/50Hz
$52,389.00USD / Each
Julabo F95-SL 22L Refrigerated/Heating Circulator, 0°C; 400V/50Hz EW-12152-03 Mfr # 9352795.07.N
COMPARE
-95 22 22L Refrigerated/Heating Circulator, 0°C; 400V/50Hz
$57,560.00USD / Each
Julabo FW95-SL 22L Water-Cooled Refrigerated/Heating Ciruclator, 0°C; 230V/60Hz EW-12152-04 Mfr # 9352796.16.N
COMPARE
-95 22 22L Water-Cooled Refrigerated/Heating Ciruclator, 0°C; 230V/60Hz
$53,296.00USD / Each
Julabo FW95-SL 22L Water-Cooled Refrigerated/Heating Ciruclator, 0°C; 400V/50Hz EW-12152-05 Mfr # 9352796.07.N
COMPARE
-95 22 22L Water-Cooled Refrigerated/Heating Ciruclator, 0°C; 400V/50Hz
$57,560.00USD / Each
Julabo FPW91-SL 22L Water-Cooled Refrigerated/Heating Ciruclator, 100°C; 230V/60Hz EW-12152-06 Mfr # 9352793.16.N
COMPARE
-91 22 22L Water-Cooled Refrigerated/Heating Ciruclator, 100°C; 230V/60Hz
$61,564.00USD / Each
Julabo FPW91-SL 22L Water-Cooled Refrigerated/Heating Ciruclator, 100°C; 400V/50Hz EW-12152-07 Mfr # 9352793.07.N
COMPARE
-91 22 22L Water-Cooled Refrigerated/Heating Ciruclator, 100°C; 400V/50Hz
$66,489.00USD / Each
Showing 1 - 20 of 34