Jenway® Quartz Cuvettes
Suitable for both UV and visible regions
  • Constructed of durable quartz
Quartz cuvettes are available in a wide capacity range from 50 µL to 35 mL and path lengths from 10 to 100 mm.
Visible % transmission: maximum from 400 to 750 nm
UV wavelength % transmission: maximum from 200 to 400 nm
$186.00 - $685.50USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Pathlength (mm) Description
Availability Pricing
COMPARE
3.5 10 Macro Cuvette, 3.5 mL, 10 mm path
$186.00USD / Each
COMPARE
14 40 Macro Cuvette, 14 mL, 40 mm path
$452.50USD / Each
COMPARE
7 20 Macro Cuvette, 7 mL, 20 mm path
$331.50USD / Each
COMPARE
0.05 10 Ultra-micro Cuvette, 50 uL, 10 mm path
$685.50USD / Each
COMPARE
0.5 10 Micro Cuvette, 500 uL, 10 mm path
$445.25USD / Each
COMPARE
1 10 Semi-micro Cuvette, 1 mL, 10 mm path
$414.00USD / Each
COMPARE
0.1 10 Ultra-micro Cuvette, 100 uL, 10 mm path
$647.50USD / Each
COMPARE
17.5 50 Macro Cuvette, 17.5 mL, 50 mm path
$423.25USD / Each
COMPARE
35 100 Macro Cuvette, 35 mL, 100 mm path
$657.50USD / Each
COMPARE
1 10 Semi-micro Cuvette, 1 mL, 10 mm path, black
$437.25USD / Each
Showing 1- 10 of 10