Ismatec® Replacement Cartridges and Adapters
Choose new cassettes based upon material of construction, tubing type and pressure capabilities
  • Material of construction standard POM-C or optional PVDF
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.
$9.40 - $153.50USD / Pkg of 4
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Housing Material Description
Availability Pricing
COMPARE
POM-C Tubing Retaining Adapters, 2-Stop, POM-C, 1/pk
$9.40USD / Pkg of 1
COMPARE
POM-C Tubing Retaining Adapters, 2-Stop, POM-C, 4/pk
$36.50USD / Pkg of 4
COMPARE
POM-C Tubing Retaining Adapters, 2-Stop, POM-C, 8/pk
$68.00USD / Pkg of 8
COMPARE
POM-C Tubing Retaining Adapters; 2-Stop, POM-C, 12/pk
$128.00USD / Pkg of 12
COMPARE
PVDF Tubing Retaining Adapters, 2-Stop, PVDF, 1/pk
$9.55USD / Pkg of 1
COMPARE
POM-C Pressure Lever Cassette / Cartridge, 3-Stop, POM-C
$85.50USD / Each
COMPARE
PVDF Pressure Lever Cassette / Cartridge, 3-Stop, PVDF
$91.50USD / Each
COMPARE
POM-C Pressure Lever Cassette / Cartridge, 2-Stop, POM-C
$73.00USD / Each
COMPARE
POM-C Click-n-go Cassette / Cartridge, 2-Stop, POM-C
$50.50USD / Each
COMPARE
POM-C Pressure Lever Cassette / Cartridge, 2-Stop, PVDF
$99.00USD / Each
COMPARE
PVDF Click-n-go Cassette / Cartridge, 3-stop, PVDF
$153.50USD / Each
COMPARE
POM-C Click-n-go Cassette / Cartridge, 3-Stop, POM-C
$64.50USD / Each
Showing 1- 12 of 12