This product is now discontinued, View Related Products
Instek APS7000 Series 500/1000 VA Programmable AC Power Sources
Electronics manufacturing and compliance testing made easy
  • Simulates abnormal line voltage, frequency, and phase fluctuations for compliance testing purposes
  • Measures RMS voltage and current, and frequency to analyze electronics characteristics
  • Commit protocols to internal memory or USB flash drive to quickly recall and initiate routine tests
  • Remotely control using USB Host or LAN
This combination power source and analyzer is ideal for electronics development, manufacturing, quality control, and IEC compliance testing. Because it measures small currents with high resolution, it is suitable for the LED industry and for testing "standby mode" power consumption.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
GW Instek APS7050 Programmable AC Power Source, 500 VA EW-20026-01 Mfr # APS7050
COMPARE
4.2 A (16.8 A peak), 2.1 A (8.4 A peak) 0 to 310.0 VAC
Call For Price
GW Instek APS7100 Programmable AC Power Source, 1000 VA EW-20026-03 Mfr # APS7100
COMPARE
8.4 A (33.6 A peak), 4.2 A (16.8 A peak) 0 to 310.0 VAC