Industrial Pressure Transmitters with Flush Diaphragm Connection
Designed for cloggong media such as slurry or motor oil applications
  • Highly resistant to pressure spikes and vibration
  • Stainless steel case and wetted parts
  • 4 to 20 mA 2-wire output signal (others available)
  • Models with cooling element allow for use in high-temperature applications
The S-11 transmitters feature a flush diaphragm process connection. They are specifically designed for the measurement of viscous fluids or media containing solids that may clog an NPT process connection. For high temperature media an integral cooling element is available. This option increases the maximum media temperature to 300°F (149°C).
Transmitters feature a printed circuit board that uses state-of-the-art surface mount technology and is potted in silicone gel for protection against mechanical shock, vibration, and moisture. Each transmitter is individually temperature compensated to assure accuracy and long-term stability even when exposed to severe ambient temperature variations.
A wide range of electrical connection and process connection options are available to meet almost any requirement. Contact an Applications Specialist for other options and configurations.
Applications
Hydraulics, pneumatics and vacuum
Test equipment
Liquid level measurement
Press control
Compressor control
Pump protection
$701.51 - $893.26USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI)
Availability Pricing
Wika 4204051 Pressure Transmitter w/ Flush Seal, 0-5 psi EW-68077-00 Mfr # 4204051
COMPARE
±0.25% BFSL 5
$701.51USD / Each
Wika 8395746 Pressure Transmitter w/ Flush Seal, 0-30 psi EW-68077-03 Mfr # 8395746
COMPARE
±0.25% BFSL 30
$701.51USD / Each
Wika 8341724 Pressure Transmitter w/ Flush Seal, 0-100 psi EW-68077-05 Mfr # 8341724
COMPARE
±0.25% BFSL 100
$701.51USD / Each
Wika 8341171 Pressure Transmitter w/ Flush Seal, 0-300 psi EW-68077-08 Mfr # 8341171
COMPARE
±0.25% BFSL 300
$701.51USD / Each
Wika 8340646 Pressure Transmitter w/ Flush Seal, 0-5000 psi EW-68077-13 Mfr # 8340646
COMPARE
±0.25% BFSL 5000
$701.51USD / Each
Wika 9749581 Pressure Transmitter w/ Flush Seal, 0-8000 psi EW-68077-14 Mfr # 9749581
COMPARE
±0.25% BFSL 8000
$701.52USD / Each
Wika 50996216 Pressure Transmitter w/ Flush Seal and Cooling, 0-200 psi EW-68077-24 Mfr # 50996216
COMPARE
±0.25% BFSL 200
$893.26USD / Each
Wika 4374394 Pressure Transmitter w/ Flush Seal and Cooling, 0-300 psi EW-68077-25 Mfr # 4374394
COMPARE
±0.25% BFSL 300
$820.14USD / Each
Wika 50716212 Pressure Transmitter w/ Flush Seal and Cooling, 0-1000 psi EW-68077-27 Mfr # 50716212
COMPARE
±0.25% BFSL 1000
$820.14USD / Each
Showing 1- 9 of 9