Implen NanoPhotometer UV/Vis Spectrophotometers
Microvolume measurement with on-the-go analysis
  • Flexible control and data collection via computer, Mac, smartphone, or tablet
  • Choose a Touch model for a built-in touch screen tablet
  • Slot for a Kensington™ lock provides antitheft security
For those in need of flexibility, this family of UV/visible spectrophotometers offers reliable protein and DNA measurements for mobile and laboratory applications. Requiring only a minute sample size of 0.3 µL and with a limit of detection down to 0.1 ng/µL (ds DNA) or 0.003 mg/µL (BSA), this spectrophotometer series accommodates any application. Simplified optics with two precisely defined pathlengths provide two fixed and sealed anchor points, prohibiting drift and eliminating the need for recalibration.
Equipped with Wi-Fi, USB A/B, HDMI and LAN interface connections for flexible instrument control and data saving options. Front USB flash drive port offers quick and easy data transfer.
$9,186.00 - $10,817.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth Max Wavelength (nm) Min Wavelength (nm)
Availability Pricing
Implen N50 NanoPhotometer UV/Vis Spectrophotometer with Touch Screen, 110/220 V EW-83070-56 Mfr # N50-Touch
COMPARE
< 5 nm 650 200
$10,817.00USD / Each
Implen C40 NanoPhotometer UV/Vis Spectrophotometer Mobile System, 110/220 V EW-83070-62 Mfr # C40-Mobile
COMPARE
< 1.8 nm 900 200
$9,186.00USD / Each