IKA RW 20 Digital Dual-Range Mixers
Two speed ranges to suit a variety of mixing applications
  • Digital display makes reproducibility easy
  • Through-shaft design provides virtually limitless adjustment for impeller depth
  • Maintenance-free motor rated for continuous duty
Use this versatile, affordable mixer for liquid volumes up to five gallons and viscosities up to 10,000 cps (approximately the viscosity of honey). Select speed from 60 to 2000 rpm in two ranges for a variety of applications. Through-shaft design permits adjustment of shaft length and impeller depth—allows you to place or remove mixing vessels without repositioning the mixer.
The motor and drive unit are conveniently integrated into a compact aluminum and thermoplastic housing with easy-to-use controls. The quiet, fan-cooled motor features thermal overload shut-off and an efficient heat dissipating design to ensure a long service life. Motor is maintenance-free and rated for continuous duty. Mixer features stepless speed control. The 1⁄2-inch LCD shows rotational speed in rpm and enables you to precisely control and repeat process parameters.
$1,345.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
IKA RW 20 Digital Dual-Range Mixer, 60 to 500/240 to 2000, 115 VAC EW-50705-05 Mfr # 3593001
COMPARE
115
$1,345.00USD / Each
IKA RW 20 Digital Dual-Range Mixer, 60 to 500/240 to 2000, 230 VAC EW-50705-15 Mfr # 0003593000
COMPARE
230
$1,345.00USD / Each