IKA EUROSTAR 60 Control Digital Mixer with Wireless Remote
Wireless controller lets you control and monitor all functions remotely
  • EUROSTAR 60 Control handles volumes up to 10.6 gallons (40 liters) and viscosities up to 50,000 cps
  • Ultra-clear, high-contrast LCD controller screen displays all process parameters
  • Torque trend function measures viscosity changes
  • Integrated temperature measurement lets you monitor fluid temperature
These powerful and advanced mixers feature a removable wireless controller with a multilingual LCD that lets you control and monitor all mixer functions. An integrated torque trend display is provided for the measurement of viscosity changes and a 1000 Pt RTD temperature sensor input lets you monitor and display fluid temperature. Mixers also include RS-232 and USB interfaces to control and document all parameters.
Additional control features include a timer function that allows you to preset run times and an interval function that enables you to program pause/run cycles.
Mixers feature infinitely adjustable stepless speed control. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in fluid viscosity.
The EUROSTAR 60 Control also features a through-shaft design that permits adjustment of shaft length and impeller depth—allowing you to place or remove mixing vessels without repositioning the mixer.
Quiet, fan-cooled motors feature thermal overload shut-off, and an efficient heat-dissipating design to ensure a long service life. Motor is maintenance-free and rated for continuous duty. Safety circuits ensure automatic cut-off in an anti-stall or overload condition.
$2,705.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
IKA Eurostar 60 Digital Mixer w/ Wireless Remote, 30-2000 rpm, 115VAC EW-50705-30 Mfr # 4440001
COMPARE
2000 85 115
$2,705.00USD / Each
IKA Eurostar 60 Digital Mixer w/ Wireless Remote, 30-2000 rpm, 230VAC EW-50705-35 Mfr # 0004440000
COMPARE
2000 85 230
$2,705.00USD / Each