ICI Quick Ship Cylinder Restraints
Securely restrain gas cylinders
Safely secure your cylinders with either a wall mounted or free-standing cylinder restraint.
The wall mounted cylinder restraint is constructed of cast aluminum and includes mounting hardware and nylon strap. Several units can be placed adjacent to each other to secure a group of cylinders.
The free-standing cylinder restraints are constructed from 2” x 2” welded tubular steel and can hold anywhere from 2 to 9 cylinders. Security chains provide proper restraint of cylinders.
$329.00 - $5,365.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Tubes Held Description
Availability Pricing
ICI Quick Ship Wall mounted Cylinder Restraint, for Cylinder 4" to 14" dia EW-78986-22 Mfr # CPCRWM
COMPARE
Aluminum 1 Wall mounted Cylinder Restraint, for Cylinder 4\" to 14\" dia
$329.00USD / Each
ICI Quick Ship Free Standing Cylinder Restraints, 30" x 16" x 30", Holds 2  Cylinders EW-78986-23 Mfr # CPCRFM2X1
COMPARE
Steel 2 Free Standing Cylinder Restraints, 30\" x 16\" x 30\", Holds 2 Cylinders
$1,905.00USD / Each
ICI Quick Ship Free Standing  Cylinder Restraints, 30" x 32" x 30", Holds 4  Cylinders EW-78986-24 Mfr # CPCRFM2X2
COMPARE
Steel 4 Free Standing Cylinder Restraints, 30\" x 32\" x 30\", Holds 4 Cylinders
$3,085.00USD / Each
ICI Quick Ship Free Standing  Cylinder Restraints, 30" x 48" x 30", Holds 6  Cylinders EW-78986-25 Mfr # CPCRFM2X3
COMPARE
Steel 6 Free Standing Cylinder Restraints, 30\" x 48\" x 30\", Holds 6 Cylinders
$4,270.00USD / Each
ICI Quick Ship Free Standing  Cylinder Restraints, 45" x 16" x 30", Holds 3  Cylinders EW-78986-26 Mfr # CPCRFM3X1
COMPARE
Steel 3 Free Standing Cylinder Restraints, 45\" x 16\" x 30\", Holds 3 Cylinders
$2,400.00USD / Each
ICI Quick Ship Free Standing  Cylinder Restraints, 45" x 32" x 30", Holds 6  Cylinders EW-78986-27 Mfr # CPCRFM3X2
COMPARE
Steel 6 Free Standing Cylinder Restraints, 45\" x 32\" x 30\", Holds 6 Cylinders
$3,865.00USD / Each
ICI Quick Ship Free Standing  Cylinder Restraints, 45" x 48" x 30", Holds 9  Cylinders EW-78986-28 Mfr # CPCRFM3X3
COMPARE
Steel 9 Free Standing Cylinder Restraints, 45\" x 48\" x 30\", Holds 9 Cylinders
$5,365.00USD / Each
Showing 1- 7 of 7