This product is now discontinued, View Related Products
IAC Adjustable Height Workstations
Adjustable height delivers flexability and ergonomics; accessories deliver productivity
  • Choose from basic and ESD workstations
  • Two styles are available: hydraulic hand crank and leg adjustable
  • All accessories can be used for either style workstation
Increase your productivity with these top-quality workstations. Choose from basic or ESD laminate, and hydraulic hand crank or adjustable leg. Worksurface laminate and industrial-grade particleboard is 1 ¼” thick, and has 180° rolled front edges. The worksurface is impact-resistant, and the ESD workstation is supplied with an ESD grounding kit. For workstations, see table below. The leg adjustable workstation comes in three sizes (see below) and worksurface height adjusts from 30” to 36” high in 2” increments. Adjustable Leg Extender Kit included, and leg extensions accepts floor glides. Heavy gauge support beams allow 1000 pounds distributed weight. The hydraulic hand crank workstation features cantilever leg design for unobstructed movement and plenty of legroom. All-welded heavy gauge steel supports up to 600 pounds static weight. The hand crank allows users to quickly and easily adjust the height of the worksurface a full 12”, from 30” to 42”, or any height in between! Choose between basic accessories such as shelves and drawers or Multi-Task System accessories such as tool trolley, multi bin panel, or CRT platform. MTS makes a great addition to existing workbenches-upgrade! Accessories even include air system for pneumatic tools. Accessories can be used for both styles of workstation, no drilling necessary, and are all listed below. Accessories can be used to upgrade your current workstations too!
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Height (in) Width (in) Description
Availability Pricing
IAC Industries QS-1000002-D Adjustable leg workstation, standard laminate surface, 30" x 60" EW-47545-40 Mfr # QS-1000002-D
COMPARE
30 60 Adjustable leg workstation, standard laminate surface, 30\" x 60\"
Call For Price
IAC Industries QS-1000003-D Adjustable leg workstation, standard laminate surface, 30" x 72" EW-47545-42 Mfr # QS-1000003-D
COMPARE
30 72 Adjustable leg workstation, standard laminate surface, 30\" x 72\"
Call For Price
IAC Industries QS-1000013-D Adjustable leg workstation, standard laminate surface, 36" x 72" EW-47545-44 Mfr # QS-1000013-D
COMPARE
36 72 Adjustable leg workstation, standard laminate surface, 36\" x 72\"
Call For Price
IAC Industries QS-1006002-D Adjustable leg workstation, ESD laminate surface, 30" x 60" EW-47545-46 Mfr # QS-1006002-D
COMPARE
30 60 Adjustable leg workstation, ESD laminate surface, 30\" x 60\"
Call For Price
IAC Industries QS-1006003-D Adjustable leg workstation, ESD laminate surface, 30" x 72" EW-47545-48 Mfr # QS-1006003-D
COMPARE
30 72 Adjustable leg workstation, ESD laminate surface, 30\" x 72\"
Call For Price
IAC Industries QS-1006013-D Adjustable leg workstation, ESD laminate surface, 36" x 72" EW-47545-50 Mfr # QS-1006013-D
COMPARE
36 72 Adjustable leg workstation, ESD laminate surface, 36\" x 72\"
Call For Price
IAC Industries QS-1005902-D Hydraulic hand crank workstation, accepts 60"W accessories EW-47545-80 Mfr # QS-1005902-D
COMPARE
30 60 Hydraulic hand crank workstation, accepts 60\"W accessories
Call For Price
IAC Industries 1011902-D Hydraulic hand crank workstation, ESD laminate, accepts 60"W accessories EW-47545-82 Mfr # 11011902
COMPARE
30 60 Hydraulic hand crank workstation, ESD laminate, accepts 60\"W accessories
Call For Price
Showing 1- 8 of 8