High-Tech Conversions Presaturated Sterile Cleanroom Wipes
Clean-cut edges eliminate fiber contamination
  • Choose ISO Class 3 or Class 5 compliance
  • Exceeds USP 797 and 800 requirements for critical contamination control
  • Lot numbered for traceability
  • Cobalt-60 gamma irradiated
Presaturated with 70% isopropyl alcohol (IPA) and 30% deionized (DI) water, these sterile wipes feature four clean-cut edges to eliminate fiber contamination. All wipes are gamma irradiated with Cobalt 60 and validated to Sterility Assurance Level (SAL) of 10-6 in accordance with standards and recommended practices of the ANSI/AAMI/ISO 11137:2006 guidelines. Each bag of wipes is identified with a lot number for traceability and a Certificate of Irradiation is included with every case. The wipes are available in a range of sizes and materials.
$266.49 - $335.61USD / Case of 300
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compliance Material Width (in)
Availability Pricing
High-Tech Conversions Cleanroom wipes, sterile, pre-saturated in 70% IPA, polyester, 9" x 9", 300/CS EW-15981-06 Mfr # FS-ULT70-99CB
COMPARE
ISO Class 3 Sealed-edge polyester 9
$266.49USD / Case of 300
High-Tech Conversions Cleanroom wipes, sterile, pre-saturated in 70% IPA, polycellulose, 9" x 12", 120/CS EW-15981-08 Mfr # FS-NT1-912CB
COMPARE
ISO Class 5 Polyester cellulose blend 12
$335.61USD / Case of 1400