High-Capacity One-Tier Laboratory Jar Mills
Grind more simultaneously
  • Single tier units hold two jars simultaneously
  • Heavy-duty construction holds up to 75 lb per tier
  • Grinds wet and dry materials effectively
These jar mills handle twice as much as other jar mills. The adjustable rollers accommodate jars from 2" to 10" in diameter. Milling jars and grinding media sold separately.
$3,720.00 - $4,790.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Speed (rpm) Description
Availability Pricing
COMPARE
115 20 to 300 One-tier jar high-capacity laboratory jar mill; 13\" roller, 115 VAC/60 Hz
$3,730.00USD / Each
COMPARE
230 20 to 300 One-tier jar high-capacity laboratory jar mill; 13\" roller, 230 VAC/50 Hz
$3,720.00USD / Each
COMPARE
115 20 to 300 One-tier jar high-capacity laboratory jar mill; 24\" roller, 115 VAC/60 Hz
$4,790.00USD / Each