HF Acid Eater
Neutralizes Hydrofluoric Acid into safe Organic salts
Unlike caustic soda and soda ash, which forms poisonous bi-fluoride, HF Acid Eater completely destroys the hydrofluoric acid polymer. This neutralizer has a 1:1 neutralization ratio as apposed to the 12:1 ratio of caustic soda. pH indicator additive eliminates the need for additional pH strips taking out the guess work. HF Acid Eater is nonhazardous and noncorrosive, and is excellent for a variety of uses and applications. Apply on horizontal, vertical, and perforated surfaces. Also neutralizes vapors and reduces odors.
$447.00USD / Case of 2
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Clift Industries 2900-025 HF Acid Eater Neutralizer, 2.5 Gallon; 2 Packs/Case EW-86295-03 Mfr # 2900-025
COMPARE
HF Acid Eater Neutralizer Kit, 2.5 Gallon; 2 Packs/Case
$447.00USD / Case of 2