Hemco Belt Drive Fume Hood Blowers
Superior chemical resistance for your toughest fumes
  • Epoxy coated steel for chemical resistance
  • Configured to fit in most ducting setups
  • Save space and create efficient flow by mounting on roof
These blowers are perfect for use in corrosive conditions. All blowers and impellers are epoxy coated to remove fumes efficiently. Seven discharge positions allows the blowers to suit any installation configuration.
$4,611.00 - $6,965.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Blower (hp) Exhaust Volume (CFM) Description
Availability Pricing
HEMCO Belt Drive Exhaust Blower for 36" Island Canopy Hood, 474 CFM; 115 VAC EW-33706-68 Mfr # 51705
COMPARE
1/2 476 Belt Drive Exhaust Blower for 36\" Island Canopy Hood; 115 VAC
$4,611.00USD / Each
HEMCO Belt Drive Exhaust Blower for 36" Wall Canopy Hood, 652 CFM; 115 VAC EW-33706-69 Mfr # 51709
COMPARE
1/2 652 Belt Drive Exhaust Blower for 36\" Wall Canopy Hood; 115 VAC
$5,982.00USD / Each
HEMCO Belt Drive Exhaust Blower for 72" Canopy Hoods, 1506 CFM; 115 VAC EW-33706-71 Mfr # 51731
COMPARE
1/2 1506 Belt Drive Exhaust Blower for 72\" Canopy Hoods; 115 VAC
$6,965.00USD / Each
HEMCO Belt Drive Exhaust Blower for 96" Canopy Hoods, 1771 CFM; 115 VAC EW-33706-72 Mfr # 51734
COMPARE
1/2 1771 Belt Drive Exhaust Blower for 96\" Canopy Hoods; 115 VAC
$6,965.00USD / Each