Heathrow Scientific DuraPorter™ Transport Carriers
Safely transport your biological and clinical samples
  • Silicone seal and lid clasps creates a secure tight closure
  • Lid opens a full 180° for full access to contents and easy cleaning
  • Available in three colors for easy identification
Constructed of durable, impact-resistant polycarbonate (PC), the DuraPorter carrier can safely transport biological and clinical samples from one location to the next. It can accommodate 13- and 16-mm tubes in 36- or 72-place tube rack or be used as one large storage space for larger samples or supplies. Insert separators (included) can be used to create smaller inner compartments.
The silicone watertight seal and three lid clasps provide a secure closure, keeping products and instruments inside clean and dry. Available in three colors for user identification, the transparent carrier enables you to visibly check sample integrity without opening the unit. Fold-down handle allows the carrier to be stackable. For samples that need to stay chilled, the entire carrier can be stored in a freezer or refrigerator. The carrier is autoclavable.
$136.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Overall Length (in) Description
Availability Pricing
Heathrow Scientific DuraPorter™ Sample Carrier, clear polycarbonate, 1/ea EW-06696-13 Mfr # HS120052
COMPARE
Clear with blue handle 15 Sample Carrier, clear polycarbonate, 1/ea
$136.00USD / Each
Heathrow Scientific DuraPorter™ Sample Carrier, transparent red polycarbonate, 1/ea EW-06696-14 Mfr # HS120077
COMPARE
Red with red handle 15 Sample Carrier, transparent red polycarbonate, 1/ea
$136.00USD / Each
Heathrow Scientific DuraPorter™ Sample Carrier, transparent yellow polycarbonate, 1/ea EW-06696-15 Mfr # HS120078
COMPARE
Yellow with yellow handle 15 Sample Carrier, transparent yellow polycarbonate, 1/ea
$136.00USD / Each