Hayward® Simplex Basket Strainers, PVC
Ideal product for the removal of crude solids
  • Low pressure drop with large straining capacity
  • Basket easily lifts out to clean or replace
  • True union connections permit easy removal of strainer assesmbly from process line
Protect pipes and process equipment from dirt and debris. The perforated baskets catch unwanted particles as the fluid passes through the strainer. Suitable for use in industries processing water, oil, paint, chemicals, or food.
Strainers are made of PVC and include FPM seals. The 1⁄2", 3⁄4", and 1" strainers include a 1⁄32" perforated basket; the 1-1⁄2" to 4" strainers include a 1⁄8" perforated basket. Extra baskets may be ordered separately below.
$194.00 - $1,764.00USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
Hayward SB10050ST PVC Simplex Basket Strainer; 1/2", Threaded Connection EW-29526-00 Mfr # SB1050ST
COMPARE
PVC, FPM PVC Simplex Basket Strainer; 1/2", Threaded Connection
$462.00USD / Each
Hayward SB1075ST PVC Simplex Basket Strainer; 3/4", Threaded Connection EW-29526-02 Mfr # SB1075ST
COMPARE
PVC, FPM PVC Simplex Basket Strainer; 3/4", Threaded Connection
$449.00USD / Each
Hayward SB1100ST PVC Simplex Basket Strainer; 1", Threaded Connection EW-29526-04 Mfr # SB1100ST
COMPARE
PVC, FPM PVC Simplex Basket Strainer; 1", Threaded Connection
$450.00USD / Each
Hayward SB1150ST PVC Simplex Basket Strainer; 1-1/2", Threaded Connection EW-29526-06 Mfr # SB1150ST
COMPARE
PVC, FPM PVC Simplex Basket Strainer; 1-1/2", Threaded Connection
$767.00USD / Each
Hayward SB1200ST PVC Simplex Basket Strainer; 2", Threaded Connection EW-29526-08 Mfr # SB1200ST18
COMPARE
PVC, FPM PVC Simplex Basket Strainer; 2", Threaded Connection
$767.00USD / Each
Hayward SB1300T PVC Simplex Basket Strainer; 3", Threaded Connection EW-29526-10 Mfr # SB1300T
COMPARE
PVC, FPM PVC Simplex Basket Strainer; 3", Threaded Connection
$1,764.00USD / Each
Hayward SB1400T PVC Simplex Basket Strainer; 4", Threaded Connection EW-29526-12 Mfr # SB1400T
COMPARE
PVC, FPM PVC Simplex Basket Strainer; 4", Threaded Connection
$1,764.00USD / Each
Hayward BS1100 1/32 PVC Simplex Basket f/ Strainers 1" or smaller; 1/32" perforation EW-29526-40 Mfr # BS1100 1/32
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 1" or smaller; 1/32" perforation
$194.00USD / Each
Hayward BS1100 1/16 PVC Simplex Basket f/ Strainers 1" or smaller; 1/16" perforation EW-29526-42 Mfr # BS1100 1/16
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 1" or smaller; 1/16" perforation
$200.00USD / Each
Hayward BS1200 1/32 PVC Simplex Basket f/ Strainers 1.5"-2"; 1/32" perforation EW-29526-44 Mfr # BS1200 1/32
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 1.5"-2"; 1/32" perforation
$282.00USD / Each
Hayward BS1200 1/16 PVC Simplex Basket f/ Strainers 1.5"-2"; 1/16" perforation EW-29526-46 Mfr # BS1200 1/16
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 1.5"-2"; 1/16" perforation
$283.00USD / Each
Hayward BS1200 1/8 PVC Simplex Basket f/ Strainers 1.5"-2"; 1/8" perforation EW-29526-48 Mfr # BS1200 1/8
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 1.5"-2"; 1/8" perforation
$274.00USD / Each
Hayward BS1200 3/16 PVC Simplex Basket f/ Strainers 1.5"-2"; 3/16" perforation EW-29526-50 Mfr # BS1200 3/16
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 1.5"-2"; 3/16" perforation
$283.00USD / Each
Hayward BS1400 1/16 PVC Simplex Basket f/ Strainers 3" or greater; 1/16" perforation EW-29526-52 Mfr # BS1400 1/16
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 3" or greater; 1/16" perforation
$589.00USD / Each
Hayward BS1400 1/8 PVC Simplex Basket f/ Strainers 3" or greater; 1/8" perforation EW-29526-54 Mfr # BS1400 1/8
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 3" or greater; 1/8" perforation
$608.00USD / Each
Hayward BS1400 3/16 PVC Simplex Basket f/ Strainers 3" or greater; 3/16" perforation EW-29526-56 Mfr # BS1400 3/16
COMPARE
PVC, FPM PVC Simplex Basket f/ Strainers 3" or greater; 3/16" perforation
$589.00USD / Each
Showing 1- 16 of 16