Hayward Electrically Actuated True Union Ball Valves
Ideal for remote control of process fluids
  • Precision machined stems specifically designed for actuated valves
  • Adjustable and reversible PTFE(R) PTFE seats
  • One-piece PVC or CPVC body construction
  • Includes two Viton(R) seals for double leakproof protection
  • Remote control capabilities via an auxiliary limit switch
  • Thermal overload circuit protects the motor from burnout and overheating
  • Easy connections via a wiring terminal strip
  • Unique mounting feature allows for easy conversion to manual operation
  • Mechanical brake for precise cycle control
  • Actuator comes mounted on the valve
  • NEMA 4X polypropylene housing stands up to harsh operating conditions
Choose between PVC and CPVC valve materials for a wide range of applications.
$529.00 - $708.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cv Port Size (NPT(F)) Wetted Materials
Availability Pricing
Hayward HCTB1050STACTV-EAU29 Electrically Actuated PVC True Union Ball Valve, 1/2", 115 VAC EW-01347-00 Mfr # HCTB1050STACTV-EAU29
COMPARE
8 0.5 PVC, Viton
$529.00USD / Each
Hayward HCTB1075STACTV-EAU29 Electrically Actuated PVC True Union Ball Valve, 3/4", 120 VAC EW-01347-02 Mfr # HCTB1075STACTV-EAU29
COMPARE
15 0.75 PVC, Viton
$539.00USD / Each
Hayward HCTB1100STACTV-EAU29 Electrically Actuated PVC True Union Ball Valve, 1", 115 VAC EW-01347-04 Mfr # HCTB1100STACTV-EAU29
COMPARE
29 1 , Viton
$567.00USD / Each
Hayward HCTB1150STACTV-EAU29 Electrically Actuated PVC True Union Ball Valve, 1-1/2", 120 VAC EW-01347-08 Mfr # HCTB1150STACTV-EAU29
COMPARE
90 1.5 PVC, Viton
$579.00USD / Each
Hayward HCTB1200STACTV-EAU29 Electrically Actuated PVC True Union Ball Valve, 2", 115 VAC EW-01347-10 Mfr # HCTB1200STACTV-EAU29
COMPARE
140 2 PVC, Viton
$625.00USD / Each
Hayward HCTB2050STACTV-EAU29 Electrically Actuated CPVC True Union Ball Valve, 1/2", 115 VAC EW-01347-20 Mfr # HCTB2050STACTV-EAU29
COMPARE
8 0.5 CPVC, Viton
$568.00USD / Each
Hayward HCTB2075STACTV-EAU29 Electrically Actuated CPVC True Union Ball Valve, 3/4", 115 VAC EW-01347-22 Mfr # HCTB2075STACTV-EAU29
COMPARE
15 0.75 CPVC, Viton
$580.00USD / Each
Hayward HCTB2200STACTV-EAU29 Electrically Actuated CPVC True Union Ball Valve, 2", 120 VAC EW-01347-30 Mfr # HCTB2200STACTV-EAU29
COMPARE
140 2 CPVC, Viton
$708.00USD / Each
Showing 1- 8 of 8