Hayward Diaphragm Valves with True Union Connections, PVC

Designed for use in corrosive environments
  • Multi-turn handle gives you precise control
  • Position indicator shows exact location of valve
  • Self draining allows for little dead volume in valve
  • PVC body and EPDM diaphragm
  • Tru-union connections
$137.00 - $349.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Outlet Port
Availability Pricing
Hayward DAB1005UEE Diaphragm Valve, PVC, 1/2" True Union Connection EW-07390-47 Mfr # DAB1005UEE
COMPARE
3.5 1/2" true union socket 1/2" true union socket
$137.00USD / Each
Hayward DAB1007UEE Diaphragm Valve, PVC, 3/4" True Union Connection EW-07390-49 Mfr # DAB1007UEE
COMPARE
7.5 3/4" true union socket 3/4" true union socket
$155.00USD / Each
Hayward DAB1010UEE Diaphragm Valve, PVC, 1" True Union Connection EW-07390-51 Mfr # DAB1010UEE
COMPARE
13 1" true union socket 1" true union socket
$193.00USD / Each
Hayward DAB1015UEE Diaphragm Valve, PVC, 1 1/2" True Union Connection EW-07390-53 Mfr # DAB1015UEE
COMPARE
33 1-1/2" true union socket 1-1/2" true union socket
$306.00USD / Each
Hayward DAB1020UEE Diaphragm Valve, PVC, 2" True Union Connection EW-07390-55 Mfr # DAB1020UEE
COMPARE
47 2" true union socket 2" true union socket
$349.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat