Hamilton® Gastight® Syringes with Removable Needle
Gas- and liquid-tight up to 200 psi!
  • Syringes handle corrosive gases and liquids, radioactive materials, and sterile solutions—ideal for gas and liquid chromatography
  • Sample loss is negligible at pressures up to 200 psi
Hamilton® Gastight® syringes with removable needle use a threaded hub and PTFE sealing ferrule that can be tightened to compensate for wear. The syringes include a removable stainless steel needle, which is ideal when there is a risk of clogged needle.
$84.80 - $152.00USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Needle Tip Style Syringe Capacity (µL)
Availability Pricing
Hamilton Gastight® Syringe, Small Hub Removable Needle, Beveled Tip, 10 µL, 26s G EW-07939-10 Mfr # 80030
COMPARE
26s Sharp, beveled, curved, non-coring 10
$87.40USD / Each
Hamilton Gastight® Syringe, Small Hub Removable Needle, Beveled Tip, 25 µL, 22s G EW-07939-11 Mfr # 80230
COMPARE
22s Sharp, beveled, curved, non-coring 25
$92.60USD / Each
Hamilton Gastight® Syringe, Small Hub Removable Needle, Beveled Tip, 50 µL, 22s G EW-07939-12 Mfr # 80930
COMPARE
22s Sharp, beveled, curved, non-coring 50
$103.00USD / Each
Hamilton Gastight® Syringe, Small Hub Removable Needle, Beveled Tip, 100 µL, 22s G EW-07939-13 Mfr # 81030
COMPARE
22s Sharp, beveled, curved, non-coring 100
$102.00USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Beveled Tip, 250 µL, 22s G EW-07939-14 Mfr # 81130
COMPARE
22s Sharp, beveled, curved, non-coring 250
$103.00USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Beveled Tip, 500 µL, 22 G EW-07939-15 Mfr # 81230
COMPARE
22 Sharp, beveled, curved, non-coring 500
$102.00USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Beveled Tip, 1 mL, 22 G EW-07939-60 Mfr # 81330
COMPARE
22 Sharp, beveled, curved, non-coring 1000
$126.00USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Beveled Tip, 2.5 mL, 22 G EW-07939-62 Mfr # 81430
COMPARE
22 Sharp, beveled, curved, non-coring 2500
$121.75USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Beveled Tip, 5 mL, 22 G EW-07939-63 Mfr # 81530
COMPARE
22 Sharp, beveled, curved, non-coring 5000
$136.25USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Beveled Tip, 10 mL, 22 G EW-07939-64 Mfr # 81630
COMPARE
22 Sharp, beveled, curved, non-coring 10000
$152.00USD / Each
Hamilton Gastight® Syringe, Small Hub Removable Needle, Blunt Tip, 10 µL, 22s G EW-07940-07 Mfr # 80065
COMPARE
22s 90° blunt end 10
$84.80USD / Each
Hamilton Gastight® Syringe, Small Hub Removable Needle, Blunt Tip, 25 µL, 22s G EW-07940-15 Mfr # 80265
COMPARE
22s 90° blunt end 25
$86.90USD / Each
Hamilton Gastight® Syringe, Small Hub Removable Needle, Blunt Tip, 50 µL, 22s G EW-07940-25 Mfr # 80965
COMPARE
22s 90° blunt end 50
$89.00USD / Each
Hamilton Gastight® Syringe, Small Hub Removable Needle, Blunt Tip, 100 µL, 22s G EW-07940-35 Mfr # 81065
COMPARE
22s 90° blunt end 100
$90.00USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Blunt Tip, 250 µL, 22 G EW-07940-45 Mfr # 81165
COMPARE
22 90° blunt end 250
$101.00USD / Each
Hamilton Gastight® Syringe, Large Hub Removable Needle, Blunt Tip, 500 µL, 22 G EW-07940-57 Mfr # 81265
COMPARE
22 90° blunt end 500
$101.00USD / Each
Showing 1- 16 of 16