Hamilton® Gastight® SampleLock™ Syringe
Gastight sample locking up to 500 psi
  • Gastight syringes feature an on/off valve needle
The Hamilton® Gastight® SampleLock™ syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination. An easy-to-use twist valve and a positive rear stop on SampleLock prevents loss of sample and plunger blowout.
$160.25USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Needle Tip Style Syringe Capacity (µL)
Availability Pricing
Hamilton Gastight® SampleLock™ Syringe, Large Hub Removable Needle, Beveled Tip, 50 µL, 22s G EW-07940-23 Mfr # 80956
COMPARE
22s Sharp, beveled, curved, non-coring 50
$160.25USD / Each
Hamilton Gastight® SampleLock™ Syringe, Large Hub Removable Needle, Beveled Tip, 100 µL, 22s G EW-07940-33 Mfr # 81056
COMPARE
22s Sharp, beveled, curved, non-coring 100
$160.25USD / Each
Hamilton Gastight® SampleLock™ Syringe, Large Hub Removable Needle, Beveled Tip, 250 µL, 22s G EW-07940-43 Mfr # 81156
COMPARE
22s Sharp, beveled, curved, non-coring 250
$160.25USD / Each
Hamilton Gastight® SampleLock™ Syringe, Large Hub Removable Needle, Beveled Tip, 500 µL, 22 G EW-07940-55 Mfr # 81256
COMPARE
22 Sharp, beveled, curved, non-coring 500
$160.25USD / Each