Hach HQ2100 Series Portable pH, Conductivity, TDS, Salinity, DO, and ORP Meters, 1 Channel
A robust and intuitive multiparameter meter
  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 10,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
Hach portable HQ meters offer a versatile range of measurements, including pH, conductivity, total dissolved solids (TDS), salinity, dissolved oxygen (DO), oxidation reaction potential (ORP), and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.
The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.
$1,180.00 - $2,830.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach HQ2100 Portable Multiparameter pH/Conductivity/TDS/Salinity/DO/ORP, 1 Channel EW-59005-11 Mfr # LEV015.53.2100A
COMPARE
Portable Multi-Meter pH/Conductivity/TDS/Salinity/DO/ORP, 1 Channel
$1,180.00USD / Each
Hach HQ2100 Portable Multiparameter with Conductivity Electrode, 1 m Cable EW-59005-12 Mfr # LEV015.53.21006
COMPARE
Portable Multi-Meter with Conductivity Electrode, 1 m Cable
$2,181.00USD / Each
Hach HQ2100 Portable Multiparameter with Conductivity Electrode, 5 m Rugged Cable EW-59005-13 Mfr # LEV015.53.21007
COMPARE
Portable Multi-Meter with Conductivity Electrode, 5 m Rugged Cable
$2,707.00USD / Each
Hach HQ2100 Portable Multiparameter with Dissolved Oxygen Electrode, 1 m Cable EW-59005-14 Mfr # LEV015.53.21004
COMPARE
Portable Multi-Meter with Dissolved Oxygen Electrode, 1 m Cable
$2,636.00USD / Each
Hach HQ2100 Portable Multiparameter with Dissolved Oxygen Electrode, 5 m Rugged Cable EW-59005-15 Mfr # LEV015.53.21005
COMPARE
Portable Multi-Meter with Dissolved Oxygen Electrode, 5 m Rugged Cable
$2,830.00USD / Each
Hach HQ2100 Portable Multiparameter with General-Purpose Gel pH Electrode, 1 m Cable EW-59005-16 Mfr # LEV015.53.21001
COMPARE
Portable Multi-Meter with General Purpose Gel pH Electrode, 1 m Cable
$1,905.00USD / Each
Hach HQ2100 Portable Multiparameter with Low-Maintenance Gel pH Electrode, 1 m Cable EW-59005-17 Mfr # LEV015.53.21002
COMPARE
Portable Multi-Meter with Low Maintenance Gel pH Electrode, 1 m Cable
$1,964.00USD / Each
Hach HQ2100 Portable Multiparameter with Low-Maintenance Gel pH Electrode, 5 m Rugged Cable EW-59005-18 Mfr # LEV015.53.21003
COMPARE
Portable Multi-Meter with Low Maintenance Gel pH Electrode, 5 m Rugged Cable
$2,513.00USD / Each
Showing 1- 8 of 8