GW Instek PSU Series Programmable Switching DC Power Supplies
Easily mount units into racks and cabinets
  • Overvoltage and overcurrent protection prevents DUT damage
  • Connect up to two units in series or four in parallel for higher voltage or higher output
  • USB host/device, LAN, RS-232, RS-485 and analog control interfaces standard
  • Optional GPIB interface provides added connectivity flexibility
  • Adjustable slew rate allows easy transition control of current and voltage
  • High efficiency and high power density
  • 19" Rack-mount size design
$2,368.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek PSU 6-200 DC Power Supply, 6 V, 200 A, 1200 W EW-20050-27 Mfr # PSU 6-200
COMPARE
DC Power Supply, 6 V, 200 A, 1200 W
$2,368.00USD / Each
GW Instek PSU 20-76 DC Power Supply, 20 V, 76 A, 1520 W EW-20050-29 Mfr # PSU 20-76
COMPARE
DC Power Supply, 20 V, 76 A, 1520 W
$2,368.00USD / Each
GW Instek PSU 40-38 DC Power Supply, 40 V, 38 A, 1520 W EW-20050-30 Mfr # PSU 40-38
COMPARE
DC Power Supply, 40 V, 38 A, 1520 W
$2,368.00USD / Each