GW Instek PEL 3000 Series Programmable DC Electronic Loads
Flexible power combinations for high-speed, versatile load simulations
  • Parallel connection of inputs provides higher capacity up to 9450 W
  • High slew rate is ideal for testing commercial and industrial servers
  • Sequence function allows you to perform highly efficient load simulations
  • Adjustable undervoltage, overcurrent, overvoltage, and overpower functions protect device under test
  • Display screen indicates elapsed time of load on and voltage at cut-off time
  • Setup memory stores up to 100 tests
  • Standard USB and RS-232 interface, with optional GPIB
$4,287.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek PEL-3111 DC Electronic Load, 1050 W EW-20050-08 Mfr # PEL-3111
COMPARE
DC Electronic Load, 1050 W
$4,287.00USD / Each