GW Instek GDS-3000 Series Digital Storage Oscilloscopes
Tackle complex measurement issues with ease
  • Visual Persistence Oscilloscope (VPO) technology—view video signals clearly and quickly
  • Split screen allows each input channel to be operated independently
  • Auto-range function displays waveform properly for tracking frequency and amplitude input signal changes
  • Display 28 simultaneous waveform measurements—grouping into amplitude, time, and delay
  • Front- and rear-panel USB host ports provide easy access of stored data
  • Incuded FreeWave remote monitoring software easily captures and displays data
These oscilloscopes are ideal for industrial, education, R&D, QC/QA, power supply and serial BUS design, system integration and debugging, maintenance and repair, and more. Units offer feature-rich functionality, an easy-to-navigate intuitive interface, and powerful software—allowing you to address complex measurements across the widest range of applications with confidence!
$3,614.00 - $4,536.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth (MHz) Channels Sample Rate (GS/s)
Availability Pricing
GW Instek GDS-3502 Oscilloscope, 2 Channel, 500 MHz EW-20050-68 Mfr # GDS-3502
COMPARE
500 2 4
$3,614.00USD / Each
GW Instek GDS-3504 Oscilloscope, 4 Channel, 500 MHz EW-20050-69 Mfr # GDS-3504
COMPARE
500 4 4
$4,536.00USD / Each