Guzzler® Portable Diaphragm Hand Pumps
Low-cost, high-flow pumping solution
  • Ideal for pumping shear-sensitive or particulate-laden fluids
  • Durable, portable pumps mount in any position
  • Available in cast aluminum or nonclogging Delrin® acetal plastic
  • 07090-09 has inlet and outlet on same side
Guzzler® hand pumps are designed for quick, easy fluid transfer.
Order service kits, noncollapsing wire-reinforced vinyl hoses, stainless steel hose clamps, and check valves separately in table.
$102.00 - $668.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate (Strokes/Gal) Max Flow Rate (GPM) Wetted Materials
Availability Pricing
Guzzler GH-0450D-161-10-5050-00 Diaphragm Hand Pump, 10 GPM; In/Out on Same Side EW-07090-09 Mfr # G450-H
COMPARE
7 10 Delrin plastic, Buna N diaphragm
$152.00USD / Each
Guzzler 400-H Diaphragm hand pump, 8.5 GPM, 7 Strokes/gallon
Always in Stock
EW-07090-10 Mfr # G400-H COLEP
COMPARE
7 8.5 Delrin plastic, Buna N diaphragm
$102.00USD / Each
Guzzler 500-H Diaphragm hand pump, 12 GPM, 5 Strokes/gallon
Always in Stock
EW-07090-11 Mfr # G500-H COLEP
COMPARE
5 12 Delrin plastic, Buna N diaphragm
$130.90USD / Each
Guzzler 2600 Diaphragm hand pump, 20 GPM, 3 Strokes/gallon EW-07090-12 Mfr # G2600
COMPARE
3 20 cast aluminum, Buna N diaphragm
$668.00USD / Each