Great Plains Industries Economical Totalizing Turbine Flowmeters
Easy-to-read LCD displays batch or cumulative totals
  • Simple, small, and sturdy
  • Mount on hose or in-line
Measure batch and cumulative totals in liquid transfer systems with these sturdy meters. Nylon meters are designed to measure water. Aluminum meters are designed to measure petroleum fuels. Meters can be used on any pump, pressure, or gravity feed systems. The batch total can be reset to measure flow for a single use. The cumulative total will automatically be reset to zero when the maximum reading of 9999 is met. Water models are nylon; fuel models are aluminum.
$229.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Max Flow Rate (GPM) Min Flow Rate (GPM)
Availability Pricing
Great Plains Industries Water Flowmeter/Totalizer, 3-30 GPM, 1" NPT(F), Nylon EW-05611-22 Mfr # 113255-4
COMPARE
water and thin fluids
$229.50USD / Each
Great Plains Industries Fuel Flowmeter/Totalizer, 3-30 GPM, 1" NPT(F), Nylon EW-05611-24 Mfr # 113255-1
COMPARE
fuel
$229.50USD / Each