Glove Bag Inflatable Glove Chambers
Excellent alternative to standard glove boxes
  • Effectively eliminate oxygen and moisture during material manipulation operations
  • Establish high-purity working atmosphere in minutes
  • Disposable chambers simplify cleanup
These durable, polyethylene glove chambers are reusable, yet priced to be disposable. Chambers are easy to use—just slide your equipment through the sleeve(s), seal, connect the gas port (accepts 3⁄8'' ID tubing) to your gas supply, and inflate. Glove bags have a 2.5 mil thickness and are made of polyethylene. Glove bags are not autoclaveable, nor are they sterile.
$114.00 - $503.00USD / Box of 6
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Chamber Depth (in) Chamber Height (in) Width of Equipment Sleeve (cm)
Availability Pricing
Glove Bag 108DX-17-17H Inflatable Glove Chamber, 11.5" Equipment Sleeve, 6/pk
Always in Stock
EW-04408-30 Mfr # 108DX-17-17H
COMPARE
17 11 29.2
$280.00USD / Box of 6
Glove Bag 108DX-27-17H Inflatable Glove Chambers, 22" Equipment Sleeve, 6/pk EW-04408-32 Mfr # 108DX-27-17H
COMPARE
17 21 55.9
$403.00USD / Box of 6
Glove Bag 108DX-27-27H Inflatable Glove Chamber, 22.5" Equipment Sleeve, 6/pk EW-04408-34 Mfr # 108DX-27-27H
COMPARE
27 15 57.2
$470.00USD / Box of 6
Glove Bag 108DX-37-27H Inflatable Glove Chamber, 33.5" Equipment Sleeve, 6/pk EW-04408-36 Mfr # 108DX-37-27H
COMPARE
27 18 85.1
$487.00USD / Box of 6
Glove Bag 108DX-37-37H Inflatable Glove Chamber, 34.5" Equipment Sleeve, 6/pk
Always in Stock
EW-04408-38 Mfr # 108DX-37-37H
COMPARE
25 37 87.6
$503.00USD / Box of 6
Glove Bag 108DSS-20-20H-6 Inflatable Chamber, Two 12" Equipment Sleeves; 6/Pk EW-04408-40 Mfr # 108DSS-20-20H-6
COMPARE
20 12 30.5
$344.00USD / Box of 6
Glove Bag 108D X-17-17HG Inflatable Glove Chamber, 10" Equipment Sleeve EW-04408-52 Mfr # 108D X-17-17HG
COMPARE
17 11 25.4
$115.00USD / Each
Glove Bag 108D X-27-17HG Inflatable Glove Chamber, 18 3/4" Equipment Sleeve
Always in Stock
EW-04408-54 Mfr # 108D X-27-17HG
COMPARE
17 21 48.3
$114.00USD / Each
Glove Bag 108D X-27-27HG Inflatable Glove Chamber, 22 1/2" Equipment Sleeve EW-04408-56 Mfr # 108D X-27-27HG
COMPARE
27 15 55.9
$123.00USD / Each
Glove Bag 108D X-37-27HG Inflatable Glove Chamber, 33 1/2" Sleeve; 27" L EW-04408-58 Mfr # 108D X-37-27HG
COMPARE
27 18 83.8
$138.00USD / Each
Glove Bag 108D X-37-37HG Inflatable Glove Chamber, 33 1/2" Sleeve; 37" L
Always in Stock
EW-04408-60 Mfr # 108D X-37-37HG
COMPARE
25 37 83.8
$137.00USD / Each
Showing 1- 11 of 11